Learn More
Sino Biological Recombinant Human DGKD Protein (Full Length, His Tag), Active
Supplier:  Sino Biological D2310H50
Encompass_Preferred
Recombinant full length human DGKD was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5367