Learn More
Medchemexpress LLC dGTP-d14 dilithium (2'-Deoxyguanosine-5'-triphosphate-d14 dilithium) | 2564-35-4 | 98.5% | 533.13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY138616S11MG
Encompass_Preferred
dGTP-d14 (2'-Deoxyguanosine-5'-triphosphate-d14) dilithium is deuterium labeled dGTP. It is a guanosine nucleotide that can be used in deoxyribonucleic acid synthesis. Guanosine nucleotides are highly susceptible to oxidative damage to 8-oxo-GDP, 8-O-dGTP, 8-O-GTP, and 8-O-dGTP.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-48921