Learn More
Medchemexpress LLC DHA Ceramide (C22:6 Ceramide) | 218608-43-6 | 98.7% | 609.96 | 1 ML

Supplier: Medchemexpress LLC HY16578110MM/1ML
DHA Ceramide (C22:6 Ceramide) is a solid, light yellow to yellow compound with a purity of 98.65%. It has a molecular weight of 609.96 and a chemical formula of C40H67NO3. This product is intended for research purposes only.
- Appearance: Solid, light yellow to yellow
- Purity: 98.65%
- Storage (solid): -20°C, protect from light, stored under nitrogen
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month; protect from light, stored under nitrogen
- Solubility: 100 mg/mL in DMSO (requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.