Learn More
Medchemexpress LLC DHH Mouse 100ug
SDP
Supplier:  Medchemexpress LLC HYP7339100UG
Encompass_Preferred
DHH Protein Mouse is a single non-glycosylated polypeptide chain Desert hedgehog protein (DHH) is a member of the Hedgehog family which encodes signaling molecules that play an important role in regulating morphogenesis
Catalog No. 50-001-80217