Learn More
Medchemexpress LLC HY-14645 10mg , (-)-DHMEQ CAS:287194-40-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1464510MG
Encompass_Preferred
HY-14645 10mg (-)-DHMEQ CAS: 287194-40-5 (-)-DHMEQ (Dehydroxymethylepoxyquinomicin) is a potent, selective and irreversible NF-κB inhibitor that covalently binds to a cysteine residue. (-)-DHMEQ inhibits nuclear translocation of NF-κB and shows anti-inflammatory and anticancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0136