Learn More
Medchemexpress LLC (-)-DHMEQ | 287194-40-5 | 99.8% | 261.23 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1464510MM/1ML
Encompass_Preferred
(-)-DHMEQ is a potent, selective, and irreversible NF-κB inhibitor that covalently binds to a cysteine residue. It inhibits nuclear translocation of NF-κB and demonstrates anti-inflammatory and anticancer activity.
  • Potent, selective, and irreversible NF-κB inhibitor
  • Covalently binds to a cysteine residue
  • Inhibits nuclear translocation of NF-κB
  • Demonstrates anti-inflammatory activity
  • Demonstrates anticancer activity
Catalog No. 50-003-63893