Learn More
TARGETMOL CHEMICALS INC DHODH-IN-12 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T110215MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DHODH-IN-12 is a leflunomide derivative and a weak dihydrorotate dehydrogenase (DHODH) inhibitor with a pKa of 5.07. purity: 100%
Catalog No. 50-258-1597