Learn More
Medchemexpress LLC Dhodh-in-23 | 1346705-53-0 | C24H21CIFNO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY153112100MG
Encompass_Preferred
DHODH-IN-23 is an orally active DHODH inhibitor that can be used for cancer research. It inhibits acute myeloid leukemia cell proliferation and induces CD11b expression in THP-1 cells. The compound has demonstrated significant antitumor activity in MV411 mouse xenograft models, both as a single agent and in combination with Cytarabine.
  • Inhibits acute myeloid leukemia cells proliferation
  • Induces CD11b expression in THP-1 cells
  • Shows antitumor activity in MV411 mouse xenograft model as a single agent
  • Shows antitumor activity in MV411 mouse xenograft model in combination with Cytarabine
Catalog No. 50-003-74029