Learn More
Medchemexpress LLC DHPS-IN-1 100mg | 2643300-54-1 | 406.66 g/mol | C17H13BrClN3O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY115712100MG
Encompass_Preferred
DHPS-IN-1 is a potent small-molecule inhibitor of deoxyhypusine synthase (DHPS) with demonstrated cellular activity against melanoma cell lines. The compound features a low nanomolar DHPS inhibitory potency and is supplied at high purity for research applications, with available solid and solution formats suited to in vitro and in vivo studies.
  • Potent DHPS inhibitor (IC50 = 0.014 μM).
  • Demonstrates cytotoxicity against melanoma cell lines.
  • High reported purity (99.78%).
  • Available in multiple pack sizes and as a 10 mM DMSO solution for convenience.
  • Good DMSO solubility (≈86.67 mg/mL) with recommended animal dosing vehicles.
  • Store at -20°C, protect from light; solutions stable at -80°C for extended storage.
Catalog No. 50-002-29369