Learn More
Eurofins Discoverx DHX9, active
Supplier:  Eurofins Discoverx 18019
Encompass
10 µg of active DHX9. C-terminal 6His and Avi-tagged recombinant, full length, human DHX9.
Catalog No. 50-004-56200