Learn More
Medchemexpress LLC Dhx9-in-6 (compound 620) | 2973401-56-6 | 98.9% | 532.99 g/mol | C23H18ClFN4O4S2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16001010MG
Encompass_Preferred
DHX9-IN-6 (Compound 620) is an ATP-dependent inhibitor of RNA helicase A (DHX9) used in cancer research and related molecular biology studies. The compound is supplied with characterization data-including purity, molecular weight, and safety documentation-to support laboratory use.
  • ATP-dependent inhibitor of RNA helicase A (DHX9).
  • Useful for cancer research and cellular target engagement studies.
  • High purity (98.9%) with accompanying certificate of analysis.
  • Molecular weight 532.99 g/mol; CAS number 2973401-56-6.
  • Available in small-scale research quantities (1 mg, 5 mg, 10 mg, 25 mg).
  • Provided with a datasheet and safety data sheet for handling guidance.
Catalog No. 50-002-85521