Learn More
Strem, An Ascensus Company CAS# 298-07-7. 500g. Di-(2-ethylhexyl)phosphoric acid (contains some mono). MFCD00009492
SDP
Supplier:  Strem, An Ascensus Company 931520500G
Encompass
CAS# 298-07-7. 500g. Di-(2-ethylhexyl)phosphoric acid (contains some mono). MFCD00009492. Molecular Weight: 322.42. Molecular Formula: [C4H9CH(C2H5)CH2O]2P(O)(OH). Color/form: colorless liq. Strem# 93-1520. http://www.strem.com/catalog/v/93-1520/
Catalog No. 50-901-14202