Learn More
Strem, An Ascensus Company CAS# 185812-86-6. 100mg. Di-mu-bromobis(tri-t-butylphosphino)dipalladium(I). MFCD04114019
SDP
Supplier:  Strem, An Ascensus Company 460355100MG
Encompass
CAS# 185812-86-6. 100mg. Di-mu-bromobis(tri-t-butylphosphino)dipalladium(I). MFCD04114019. Molecular Weight: 777.28. Molecular Formula: Pd2Br2[P(C4H9)3]2. Color/form: dark-green xtl. Strem# 46-0355. http://www.strem.com/catalog/v/46-0355/
Catalog No. 50-901-12329