Learn More
Strem, An Ascensus Company CAS# 12073-36-8. 250mg. Di-mu-chloro-dichlorobis(ethylene)diplatinum(II), min. 97% Zeise's dimer. MFCD00058713
SDP
Supplier:  Strem, An Ascensus Company 780460250MG
Encompass
CAS# 12073-36-8. 250mg. Di-mu-chloro-dichlorobis(ethylene)diplatinum(II), min. 97% Zeise's dimer. MFCD00058713. Molecular Weight: 588.10. Molecular Formula: [PtCl2(C2H4)]2. Color/form: orange xtl. Strem# 78-0460. http://www.strem.com/catalog/v/78-0460/
Catalog No. 50-901-13222