Learn More
Medchemexpress LLC Di-n-dodecyl phthalate-3,4,5,6-d4 | 2708280-31-1 | MFCD28009755 | >98.0% | 506.79 g/mol | C32H50D4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW353555S25MG
Encompass_Preferred
Di-n-dodecyl phthalate-3,4,5,6-d4 is a deuterium-labeled analog of di-n-dodecyl phthalate used as an analytical standard or tracer in research. Molecular formula C32H50D4O4 and molecular weight 506.79 g/mol. For research use only.
  • Deuterium-labeled tracer for isotopic and pharmacokinetic studies.
  • High isotopic enrichment (approximately 99 atom % D).
  • High chemical purity (≥98.0% reported).
  • Supplied as a neat liquid suitable for reference standards.
  • Recommended storage: pure form -20°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • Not validated for clinical use; intended for laboratory research only.
Catalog No. 50-002-85142