Learn More
Strem, An Ascensus Company CAS# 819-19-2. 25g. Di-t-butylphosphine, 98+%. MFCD00134241
SDP
Supplier:  Strem, An Ascensus Company 15104025G
Encompass
CAS# 819-19-2. 25g. Di-t-butylphosphine, 98+%. MFCD00134241. Molecular Weight: 146.22. Molecular Formula: [(CH3)3C]2PH. Color/form: colorless liq. Strem# 15-1040. http://www.strem.com/catalog/v/15-1040/
Catalog No. 50-901-10029