Learn More
Ubpbio Containing eight diUb linked via the N-terminal methionine (linear), K6, K11, K27, K29, K33, K48 or K63. Useful for assays assessing ubiquitin linkage-specificity
Supplier:  Ubpbio J2000
Encompass_Preferred
This kit contains 5 µg each of linear, K6, K11, K27, K29, K33, K48, and K63-linked diubiquitin, a total of eight different diubiquitin forms. It can be used for determining linkage specificity of deubiquitinating enzymes and polyubiquitin-binding proteins
Catalog No. 50-233-9318