Learn More
Selleck Chemical LLC diABZI STING agonist S8796-100MG
SDP
Supplier:  Selleck Chemical LLC S8796100MG
Encompass_Preferred
diABZI STING agonist (diABZI STING agonist-1 Compound 3 Tautomerism) is a potent non-nucleotide STING agonist and has tremendous potential to improve treatment of cancer in humans Solutions are unstable and should be fresh-prepared
Catalog No. 50-000-24491