Learn More
Selleck Chemical LLC Diacerein-50mg
SDP
Supplier:  Selleck Chemical LLC S426750MG
Encompass_Preferred
Diacerein is an inhibitor of pro-inflammatory cytokine Interleukin-1B (IL-1B) production, prescribed for osteoarthritis and chronic inflammatory arthritis.
Catalog No. 50-238-9677