Learn More
Medchemexpress LLC HY-N6692 5mg Medchemexpress, Diacetoxyscirpenol CAS:2270-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66925MG
Encompass_Preferred
Medchemexpress, HY-N6692 5mg Diacetoxyscirpenol CAS:2270-40-8 Diacetoxyscirpenol (DAS) is a trichothecene mycotoxin, a secondary metabolite product of fungi. Diacetoxyscirpenol (DAS) consumption induces haematological disorders (neutropenia, aplastic anemia) in human and animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0455