Learn More
Abcam Diallyl tetrasulfide, ROS inducer, 25MG
Supplier:  Abcam AB14360325MG
Encompass_Preferred
MW 210.4 Da, Purity >90%. JNK activator. ROS inducer. Irreversible Cdc25 isoforms A and C inhibitor. Targets tubulin. Induces G2-M arrest and apoptosis. Shows antiproliferative, cytoprotective and antioxidant effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3296