Learn More
Medchemexpress LLC Diallyl Trisulfide 100mg | 2050-87-5 | 178.34 g/mol | C6H10S3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117235100MG
Encompass_Preferred
Diallyl trisulfide is an analytical reference standard used in analytical chemistry and biomedical research. It is a trisulfide organosulfur compound (C6H10S3, CAS 2050-87-5) commonly used as a reference in HPLC, GC, and MS, and studied for anticancer, antioxidant, and antibacterial activities.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High reported purity (99.9%).
  • Available in small pack sizes for research use (e.g., 100 MG).
  • Storage in solution: -80°C (6 months) or -20°C (1 month) under nitrogen.
  • Used as a reference material in qualitative and quantitative assays.
Catalog No. 50-002-28513