Learn More
Medchemexpress LLC Diallyl Trisulfide 25mg | 2050-87-5 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11723525MG
Encompass_Preferred
Diallyl trisulfide (CAS 2050-87-5) is an organosulfur compound derived from garlic, supplied as a purified solid for laboratory research. It is used to investigate apoptosis, anticancer activity, and related cellular signaling pathways.
  • High purity suitable for analytical and biological studies.
  • Chemical formula C6H10S3; molecular weight 178.34 g/mol.
  • Reported bioactivities include anticancer, anti-inflammatory, antioxidant, and antibacterial effects.
  • Suitable for in vitro and biochemical research (research use only).
  • Available in small-mass packs convenient for bench-scale experiments.
Catalog No. 50-002-26350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.