Learn More
Medchemexpress LLC Diallyldimethylammon 1kg
SDP
Supplier:  Medchemexpress LLC HYW1064861KG
Encompass_Preferred
Diallyldimethylammonium chloride (60% in water) is a quaternary ammonium compound belonging to the class of alkylammonium salts The compound is widely used as a cationic monomer in the production of water-soluble polymers especially in the manufacture of flocculants and coagulants for water treatment processes In addition it can be used as an antimicrobial agent surfactant or adhesive in various industrial applications Its unique chemical properties make it an important ingredient in a variety of industrial processes including papermaking textiles and personal care products
Catalog No. 50-002-06676