Learn More
Medchemexpress LLC Diamfenetide | 36141-82-9 | 99.7% | 250 MG
SDP
Supplier:  Medchemexpress LLC HY119893250MG
Encompass_Preferred
Diamfenetide is a research reagent and analytical standard used to study Fasciola hepatica infections in vitro; it induces irreversible paralysis in immature and adult parasites.
  • Used in vitro to study Fasciola hepatica infections.
  • Induces irreversible paralysis of immature and adult parasites.
  • Provided as a high-purity analytical standard for research applications.
  • Molecular formula C20H24N2O5, molecular weight 372.41 g/mol.
  • Commonly supplied in small-quantity packs suitable for laboratory use.
Catalog No. 50-002-34605