Learn More
Apexbio Technology LLC Diammonium Glycyrrhizinate 79165-06-3 25mg
SDP
Supplier:  Apexbio Technology LLC C644825
Encompass_Preferred
Diammonium Glycyrrhizinate is a small-molecule compound structurally characterized as an ammonium salt of glycyrrhizic acid It is designed to modulate intracellular inflammatory signaling pathways thereby regulating inflammation-related cellular responses Diammonium Glycyrrhizinate exerts its biological activity primarily through the selective regulation of signaling pathways associated with enzyme-mediated signal transduction Experimental applications include the use of inflammation-associated cellular models to investigate pathway modulation both in vitro and in vivo Based on these pharmacological properties Diammonium Glycyrrhizinate holds research potential in elucidating therapeutic mechanisms relevant to inflammatory diseases
Catalog No. 50-000-89642