Learn More
Medchemexpress LLC Diammonium hydrogen phosphate | 7783-28-0 | MFCD00010887 | 99.1% | 132.06 g/mol | H9N2O4P | 100g
SDP
Supplier:  Medchemexpress LLC HYY0302100G
Encompass_Preferred
Diammonium phosphate is a neutral and efficient catalyst in 1 8 Dioxo octahydroxanthene derivatives synthesis Diammonium phosphate can be used as an excipient such as diuretic buffer effervescent[1][2]
Catalog No. 50-002-69809