Learn More
Electron Microscopy Sciences Optical Fiber Diamond Cleaver Tool
SDP
Supplier:  Electron Microscopy Sciences 62165DC
Encompass_Preferred
Retractable, ground and polished natural diamond which has a v90 deg included angle with a relief angle of 30 deg . Cleaver is retained in a metal pen like casing with a pocket clasp. Diamond is 1.5mm wide x 3.0mm long, overall length
Catalog No. 50-315-22