Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Diamond Grinding Disc, 75 µm, 8" diameter
SDP

Supplier:  Electron Microscopy Sciences 5034575

Encompass

Dialok Diamond Discs consists of a flat, stainless steel backing with sharp, blocky diamond particles electroformed onto the disc. The special hard nickel alloy that is used in Dialok is bonded rigidly and it locks the diamond particles in the disc thus providing fast cutting of all hard materials. The sharp particles cut cleanly with no edge rounding. The discs are supplied with pressure sensitive adhesive backing (PSA).
Particles: 75 µm
Diameter: 8"

Catalog No. 50-997-242


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.