Learn More
Electron Microscopy Sciences Diamond Lapping Film 8", PSA Backing, 9 µm Particles
SDP
Supplier:  Electron Microscopy Sciences 5035125
Encompass_Preferred
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 9 µm
Catalog No. 50-997-258