Learn More
Thomas Scientific Diamond Midi Centrifuge Tube, 5.0mL, PP, Assembled Blue Screw Cap, Graduated, STERILE, 20/Bag, 10 Bags/Unit, 200 box
SDP
Supplier:  Thomas Scientific 1139Z20
Encompass
An efficient and economical alternative to 15mL conical tubes when working with sample volumes between 2.0mL and 5.0mL.
Catalog No. NC1753171