Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Cone Square Grid, 3.8 mm Tip, 038 I.S.O.
SDP
Supplier:  Electron Microscopy Sciences 6215538
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 3.8 mm
I.S.O. size: 038
Catalog No. 50-254-46