Learn More
Ted Pella Inc DIAMOND SCRIBE 0.25MM TIP DIA.
SDP
Supplier:  Ted Pella Inc 13663
Encompass
Diamond Scribe, 0.25mm Tip dia., Positive chuck clamping, Low-friction, Bronze bushing, Non-roll handle, Knurled finger grip, Lightweight, 1/EA
Catalog No. NC9261048