Learn More
Electron Microscopy Sciences Dumont Straight Positive Action Diamond Coated Tweezers, 165 mm OAL, 0.25 mm Tip width
SDP
Supplier:  Electron Microscopy Sciences 78100
Encompass_Preferred
Dumont Stainless Steel tweezers coated with an extremely fine diamond dust in the inside tip. They are essential in applications where a tight grip is necessary. They are also available with an improved locking system. They are 5 1/2" (150 mm) long. They may not be suitable for use with EM grids.
OAL: 150 mm
Tip width: 0.25 mm
Tip thickness: 0.25 mm
Catalog No. 50-237-75