Learn More
Electron Microscopy Sciences Precision Diamond Wafer Blade 4", Coarse grit with High Concentration, 0.012" thick
SDP
Supplier:  Electron Microscopy Sciences 50266CH4
Encompass_Preferred
These bonded blades are constructed of an inner metal core and an outer rim, this rim is bonded with diamond particles. These blades come in high and low concentrations of diamond particles to handle various sectioning requirements. A high concentration of coarse diamond particles is for general laboratory use with low or high saw's speed.
Particles: Coarse
Concentration: High
Thickness: 0.012"
Catalog No. 50-997-205