Learn More
Electron Microscopy Sciences Diamond Wafer Blade 5", 0.20" W, 0.276"D
SDP
Supplier:  Electron Microscopy Sciences 5026605
Encompass_Preferred
The blades are constructed of an inner metal core and an outer rim with diamond particles. This diamond wafering blade has a diameter of 4" and a thickness of .20" RIM,20"D.
Catalog No. 50-997-202