Learn More
Alkali Scientific Diamond Weigh Boats, White, 100mL, 100 X 125mm, 250/cs
SDP
Supplier:  Alkali Scientific J6503W
Encompass_Preferred
ASI Weigh Boats are made of antistatic polystyrene and will withstand temperatures to 90˚C, as well as exposure to diluted acids, alcohols, bases and aqueous solutions. Their flexibility and smooth interior surfaces with rounded corners allow for easy decanting of powdered samples. Their flat bottoms ensure stability and prevent tipping. Weigh Boats are available in a diamond shape for easier pouring and come in white. Originally designed as weighing containers, they have proven ideal for dispensing and storage and as disposable mixing/quick freezing trays. 250/pk
Catalog No. 50-233-6167