Learn More
DSI LLC DIAMONDBOND - C18 REVERSED-PH
Supplier:  DSI LLC DB010546
Encompass_Preferred
DiamondBond-C18 eliminates the shortcomings of conventional ODS-silica columns by combining patented surface technology with ZirChrom's state-of-the-art particles. Carbon-clad zirconia, stable at all levels of pH.
Catalog No. 50-123-043