Learn More
Medchemexpress LLC Diamthazole hydrochloride | 17140-69-1 | 99.9% | 329.89 | 50 MG

Supplier: Medchemexpress LLC HY148160A50MG
Diamthazole (Dimazole) hydrochloride is an antifungal agent primarily used for the research of infection. It is presented as a solid, with a color ranging from white to light yellow. The molecular formula for Diamthazole hydrochloride is C15H24ClN3OS, and its molecular weight is 329.89.
- Purity of 99.86%.
- Shipped at room temperature within the continental US.
- Store at 4°C in a sealed container, protected from moisture. In solvent, store at -80°C for 6 months or -20°C for 1 month in sealed storage away from moisture.
- Solubility of 100 mg/mL (303.13 mM) in DMSO, achievable with ultrasonic treatment and warming.
- For research purposes only, not for human application or direct sale to patients.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.