Learn More
Medchemexpress LLC Dianhydrodulcitol | 23261-20-3 | 98.0% | 146.14 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY165135MG
Encompass_Preferred
VAL-083 is an alkylating agent that creates N7 methylation on DNA, exhibiting antitumor activity. It suppresses cell growth and induces apoptosis in U251 and SF188 cells, and inhibits T98G cell growth in a dose-dependent manner. VAL-083 also inhibits the proliferation, migration, and invasion of HUVEC and U251 cells, reducing the expression of MMP2, VEGF, VEGFR2, and FGF2.
  • Induces cell cycle arrest at the G2/M phase in glioma cell lines.
  • Enhances radiosensitivity by activating the p53-p21 and CDC25C-CDK1 cascades.
  • Inhibits angiogenesis in zebrafish.
  • Significantly blocks the growth of LN229 cells in mice.
Catalog No. 50-003-91779