Learn More
Medchemexpress LLC Diarylpropionitrile | 1428-67-7 | 99.7% | 239.27 g/mol | C15H13NO2 | 100MG
SDP
Supplier:  Medchemexpress LLC HY12452100MG
Encompass_Preferred
DPN (Diarylpropionitrile) is a non-steroidal, ERβ-selective research ligand used to probe estrogen receptor β signaling. It exhibits an in vitro EC50 of 0.85 nM and has been reported to show neuroprotective effects in preclinical studies. The product is supplied as a powder, soluble in DMSO (100 mg/mL), with recommended storage at -20°C for the solid and lower temperatures for solutions.
Catalog No. 50-002-35602