Learn More
Selleck Chemical LLC Diatrizoate sodium S4816-25mg
SDP
Supplier:  Selleck Chemical LLC S481625MG
Encompass_Preferred
Diatrizoate sodium (Amidotrizoate sodium Hypaque sodium Triombrin) is an iodinated radiopaque X-ray contrast medium that is used as a diagnostic aid in angiography urography and radiography
Catalog No. 50-311-2023