Learn More
Medchemexpress LLC Diatrizoic acid | 117-96-4 | 99.7% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB09261G
Encompass_Preferred
Diatrizoic acid is an iodinated radiocontrast agent used for radiographic imaging of the airways. It functions by inducing mitochondrial turnover, oxidative stress, and activating apoptosis through calcium dysregulation. This chemical is supplied as a solid and is stable under recommended storage conditions.
  • Iodinated radiocontrast agent
  • Used for radiographic imaging of the airways
  • Induces mitochondrial turnover and oxidative stress
  • Activates apoptosis by dysregulating calcium
  • Solid appearance
  • Stable under recommended storage conditions
Catalog No. 50-004-09082