Learn More
Medchemexpress LLC Diatrizoic acid | 117-96-4 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB092610MM/1ML
Encompass_Preferred
Diatrizoic acid (Diatrizoate) is an iodinated radiocontrast agent with potential for radiographic imaging of airways. It induces mitochondrial turnover, oxidative stress, and activates apoptosis by dysregulating calcium.
  • Iodinated radiocontrast agent
  • Potential for radiographic imaging of the airways
  • Induces mitochondrial turnover and oxidative stress
  • Activates apoptosis by dysregulating calcium
  • Nanoparticle agglomerates offer a promising radiocontrast agent for safe and effective lung visualization
Catalog No. 50-004-07695