Learn More
Medchemexpress LLC Diaveridine | 5355-16-8 | 99.8% | 1 G

Supplier: Medchemexpress LLC HYB19021G
Diaveridine (EGIS-5645) is a dihydrofolate reductase (DHFR) inhibitor and an antibacterial agent, with a Ki of 11.5 nM for the wild type DHFR. This compound is intended for research use only.
- Dihydrofolate reductase (DHFR) inhibitor
- Antibacterial agent
- Ki of 11.5 nM for the wild type DHFR
- Molecular Weight: 260.29
- Formula: C13H16N4O2
- Appearance: Solid
- Color: White to off-white
- Purity: 99.79%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.