Learn More
Selleck Chemical LLC Diaveridine S5200-1g
SDP
Supplier:  Selleck Chemical LLC S52001G
Encompass_Preferred
Diaveridine (AI3-23935 CCRIS-3784) is coccidiostat with anti-parasitic activity and also a dihydrofolate reductase (DHFR) inhibitor preventing folic acid synthesis in species of Pneumocystis
Catalog No. 50-311-1717