Learn More
Medchemexpress LLC Dibenzocyclooctyne-amine | 1255942-06-3 | MFCD22380759 | 99.1% | 276.34 | C18H16N2O | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0004231G
Encompass_Preferred
DBCO-amine is a dibenzocyclooctyne-functionalized amine reagent used for copper-free click chemistry (strain-promoted alkyne-azide cycloaddition, SPAAC). It is supplied at high purity for bioconjugation and linker synthesis, and is suitable for labeling azide-bearing biomolecules.
  • Enables copper-free SPAAC bioconjugation with azide groups
  • High purity suitable for synthetic and bioconjugation workflows
  • Available in multiple pack sizes, including 1 g for scale-up
  • Used for ADC linker synthesis and labeling applications
  • Soluble in common organic solvents such as DMSO
Catalog No. 50-002-02807