Learn More
Medchemexpress LLC Dibenzothiophene-4-boronic acid | 108847-20-7 | 228.07 | 10 G

Supplier: Medchemexpress LLC HYW00276210G
4-Dibenzothiopheneboronic acid is a biochemical reagent used as a biological material or organic compound for life science-related research. It is intended for research use only and is not sold to patients.
- Appearance: solid
- Color: white to off-white
- Formula: C12H9BO2S
- Shipping: room temperature in continental US; may vary elsewhere
- Storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.