Learn More
Medchemexpress LLC Dibenzothiophene-4-boronic acid | 108847-20-7 | 228.07 | 25 G

Supplier: Medchemexpress LLC 25GHYW002762
Dibenzothiophene-4-boronic acid is a biochemical reagent suitable for life science related research as a biological material or organic compound. This product is for research use only.
- Purity: 99.88%
- Molecular weight: 228.07
- Formula: C12H9BO2S
- CAS number: 108847-20-7
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.