Learn More
Selleck Chemical LLC Dibenzothiophene S4205-50mg
SDP
Supplier:  Selleck Chemical LLC S420550MG
Encompass_Preferred
Dibenzothiophene (DBT Diphenylene sulfide) is a model compound for organic sulfur in fossil fuels
Catalog No. 50-310-9418